missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, 5% (w/w), Ricca Chemical

Catalog No. 749716
Change view
Click to view available options
Package: Amber Glass; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
749716 Package: Amber Glass; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 749716 Supplier Ricca Chemical Company Supplier No. 749716

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7632-00-0, 7732-18-5
CAS Min % 93.0, 4.81
CAS Max % 97.0, 5.0
Molecular Formula NNaO2
MDL Number MFCD00011118
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
SMILES [Na+].[O-]N=O
IUPAC Name sodium nitrite
Molecular Weight (g/mol) 69.00
PubChem CID 23668193
ChEBI CHEBI:78870
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Concentration or Composition Notes 5% (w/w)
Chemical Name or Material Sodium Nitrite
Show More Show Less