missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-octadecyl sulfate, 98% (dry wt.), water <7%

Catalog No. AAA1504214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1504214 25 g
AAA1504203 1 g
AAA1504206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1504214 Supplier Thermo Scientific Chemicals Supplier No. A1504214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1120-04-3 | C18H37NaO4S | 372.54 g/mol

Sodium n-octadecyl sulfate is used in the formulation of hair care products including shampoo, skin cleansers, and other skin care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-octadecyl sulfate is used in the formulation of hair care products including shampoo, skin cleansers, and other skin care products.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 1120-04-3
Molecular Formula C18H37NaO4S
Molecular Weight (g/mol) 372.54
MDL Number MFCD00007469
InChI Key NWZBFJYXRGSRGD-UHFFFAOYSA-M
Synonym sodium stearyl sulfate, octadecyl sulfate sodium salt, sodium octadecyl sulfate, octadecyl sodium sulfate, sulfuric acid, monooctadecyl ester, sodium salt, sodium octadecyl sulphate, sodium monostearyl sulfate, unii-t4eww0y4ej, sodium n-octadecyl sulfate, sodium monooctadecyl sulfate
PubChem CID 23673604
IUPAC Name sodium;octadecyl sulfate
SMILES CCCCCCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 176°C to 180°C
Linear Formula CH3(CH2)17OSO3Na
Quantity 25 g
Beilstein 3640871
Solubility Information Soluble in water.
Formula Weight 372.55
Percent Purity 98%
Assay (dry wt.), water <7%
Chemical Name or Material Sodium n-octadecyl sulfate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-295-0
RTECSNumber WT1170000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only