missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methyl Sulfate 98.0+%, TCI America™

Catalog No. M043325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M043325G 25 g
M0433250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M043325G Supplier TCI America Supplier No. M043325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 512-42-5
Molecular Formula CH3NaO4S
Molecular Weight (g/mol) 134.08
MDL Number MFCD00013457
InChI Key DZXBHDRHRFLQCJ-UHFFFAOYSA-M
Synonym sodium methyl sulfate, methylsulfuric acid sodium salt, methyl sodium sulfate, sodium methyl sulphate, sodium monomethylsulfate, unii-38w76yj3jn, sodium methylsulfate, methyl sulfate sodium salt, methyl sulfate, sodium salt, sulfuric acid, monomethyl ester, sodium salt
PubChem CID 2735086
IUPAC Name sodium methyl sulfate
SMILES [Na+].COS([O-])(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 134.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Methyl Sulfate
EINECSNumber (2)-4121
TSCA Yes