missing translation for 'onlineSavingsMsg'
Learn More

Sodium methoxide, pure, titrant, (0.5 M in methanol)

CAS: 124-41-4 | CH3NaO | 54.024 g/mol

Supplier:  Thermo Scientific Chemicals 419600025

 View more versions of this product

Catalog No. AC419600025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium methoxide
0.5 M in methanol
65.0°C
CH3NaO
MFCD00012179
01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium;methanolate
10942334
Pure
0.8
11°C
Colorless
124-41-4 , 67-56-1
-98.0°C
CH3ONa
0.45 to 0.60M
0.8000g/mL
Solubility in water: reacts.
C[O-].[Na+]
54.024
54.02
Glass bottle
2.5 L
Liquid

Chemical Identifiers

124-41-4
54.024
WQDUMFSSJAZKTM-UHFFFAOYSA-N
10942334
C[O-].[Na+]
CH3NaO
MFCD00012179
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
sodium;methanolate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes serious eye irritation. Causes skin irritation. Causes damage to organs. Highly flammable liquid and vapor. Highly flammable liquid and vapor.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS Signal Word: Danger

EINECSNumber : 204-699-5

RTECSNumber : PC3570000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only