accessibility menu, dialog, popup

UserName

CAS RN 124-41-4

IUPAC Name: sodium methanolate
Synonyms: sodium methoxide
Molecular Weight (g/mol): 54.02
Molecular Formula: CH3NaO
InChi Key: WQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES: [Na+].C[O-]
Molecular Weight (g/mol) 
  • (17)
  • (13)
Quantity 
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
Grade 
  • (2)
  • (3)
  • (1)
  • (11)
  • (4)
Physical Form 
  • (2)
  • (13)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (22)
  • (3)
  • (4)
  • (23)
  • (17)
  • (13)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (11)
  • (4)
  • (2)
  • (13)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (5)