missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium methoxide, pure, titrant, (0.5 M in methanol)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 65.0°C |
| Melting Point | -98.0°C |
| Linear Formula | CH3ONa |
| Concentration | 0.45 to 0.60M |
| Fieser | 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352 |
| Density | 0.8000g/mL |
| Solubility Information | Solubility in water: reacts. |
| Formula Weight | 54.02 |
| Grade | Pure |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS P Statement Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes serious eye irritation. Causes skin irritation. Causes damage to organs. Highly flammable liquid and vapor. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. |
| EINECSNumber | 204-699-5 |
| RTECSNumber | PC3570000 |
| TSCA | TSCA |
RUO – Research Use Only