missing translation for 'onlineSavingsMsg'
Learn More

Sodium metasilicate pentahydrate, tech.

Catalog No. AA14103D9
Change view
Click to view available options
Quantity:
1 kg
5 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA14103D9 5 x 1 kg
AA14103A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA14103D9 Supplier Thermo Scientific Chemicals Supplier No. 014103D9

CAS: 10213-79-3 | H10Na2O8Si | 212.137 g/mol

Sodium metasilicate pentahydrate is used as a starting material in the preparation of silicas and zeolites. It finds application in detergents and cleaners, pulp and paper production. It is also used in soil stabilization, sealing, adhesives and binders. Further, it is used in titanium dioxide production, paints for masonry and glass, spray coatings for tunnel construction. In addition to this, it is used in fruit and vegetable washes and sanitizers for food-contact surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10213-79-3
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.137
MDL Number MFCD00149176
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate pentahydrate, silicic acid, disodium salt, pentahydrate, disodium oxosilanediolate pentahydrate, silicic acid h2sio3 , disodium salt, pentahydrate, 2na.so3i.5h2o, sodium meta-silicate pentahydrate, sodium silicate solution water glass, disodium dioxido oxo silane pentahydrate, sodium metasilicate pentahydrate, tech., disodium oxosilanebis olate pentahydrate
PubChem CID 61495
IUPAC Name disodium;dioxido(oxo)silane;pentahydrate
SMILES O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Melting Point 72°C
Color White
Physical Form Granules
Quantity 5 x 1 kg
UN Number UN3253
Merck Index 14,8642
Solubility Information Soluble in cold water.
Formula Weight 212.14 (122.06 Anhydrous)
Grade Technical
Odor Odorless
Appearance White granules
Density 1.75
Chemical Name or Material Sodium metasilicate pentahydrate
Show More Show Less
Hazard Category H290-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: DISODIUM TRIOXOSILICATE
EINECSNumber 229-912-9
RTECSNumber VV9287500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only