missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metasilicate Pentahydrate, Purified, Reagents

Catalog No. 01336977
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336977 3 kg
01336976 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336977 Supplier Labchem Inc Supplier No. C23248003A5

Sodium Metasilicate Pentahydrate, Purified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10213-79-3
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.14
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
IUPAC Name disodium oxosilanebis(olate) pentahydrate
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

Specifications

Boiling Point >1200°C
Color Colorless/White
Physical Form Solid
Quantity 3 kg
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.52
Chemical Name or Material Sodium Metasilicate Pentahydrate, Purified