missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium metasilicate nonahydrate, 44-47.5% total solids
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 72.0°C |
| Color | White |
| Physical Form | Granules or Granular Powder |
| Quantity | 500 g |
| Linear Formula | Na2SiO3·9H2O |
| Merck Index | 15, 8804 |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 284.19 |
| Percent Purity | 44 to 47.5% total solids |
| Packaging | Plastic bottle |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May be corrosive to metals. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with wat |
RUO – Research Use Only