missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Metasilicate, MP Biomedicals
Specifications
Specifications
| Quantity | 100 g |
| Assay Percent Range | ≥97% |
| Formula Weight | 284.2 |
| Grade | Reagent |
| Chemical Name or Material | Sodium Metasilicate |
Safety and Handling
| Recommended Storage | Room Temperature |