missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metabisulfite Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015470
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015470 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015470 Supplier Merck Emd Millipore Supplier No. PHR14341G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. Sodium Metabisulfite is a sulfiting agent, widely employed as antioxidants, preservatives and browning inhibitors in pharmaceutical formulations and a variety of food and beverages.

Specifications

CAS 7681-57-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula Na2S2O5
MDL Number MFCD00167602
Melting Point >300°C
Synonym Sodium metabisulfite; Sodium disulfite; Sodium pyrosulfite
Type Certified Reference Material
Quantity 1 g
Formula Weight 190.11
Grade Certified Reference Material