missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metabisulfite (Granular/NF/FCC), Fisher Chemical™

Catalog No. S242500
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S242500 500 g Poly Bottle
S242212 2.5 kg Poly Bottle
S24212 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S242500 Supplier Thermo Fisher Scientific Supplier No. FLS242 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Melting Point 150°C
Color White
pH 4 to 6
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 190.11
Percent Purity 90.0 to 100.0% (FCC)
Grade FCC/NF
Identification Pass Test (FCC), Pass Test (FCC, NF), Pass Test (NF)
Packaging Poly Bottle
Density 1.4g/cm³
Chemical Name or Material Sodium Metabisulfite
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. May cause skin and respiratory tract irritation. Contact with acids liberates toxic gas. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:0
Instability:1