missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metabisulfite, Granular, NF, EP, BP, JP, 65-67.4%, Spectrum™ Chemical

Catalog No. 18606655
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606655 45 kg Fiber Drum
18606653 2.5 kg Poly Bottle
18606654 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606655 Supplier Spectrum Chemical Mfg Cor Supplier No. SO18245KGBL

SO182, 7681-57-4, Na2S2O5

Spectrum™ Chemical Sodium Metabisulfite, Granular, NF, EP, BP, JP is commonly used as a preserving agent, antioxidant and disinfectant. All Spectrum Chemical NF, EP, BP, and JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

CAS Max % 1
Quantity 45 kg
Percent Purity 65 to 67.4%
Grade Multi-Compendial
Packaging Fiber Drum
Appearance Clear