missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metabisulfite, Granular, FCC, 90-100.5%, Spectrum™ Chemical

Catalog No. 18606651
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606651 50 kg Poly Bottle
18606648 500 g Poly Bottle
18606649 2.5 kg Poly Bottle
18606650 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606651 Supplier Spectrum Chemical Mfg Cor Supplier No. SO18150KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SO181, 7681-57-4, Na2S2O5

Spectrum™ Chemical Sodium Metabisulfite, Granular, FCC is used as a preservative and antioxidant in food and is also known as E223. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 50 kg
Percent Purity 90 to 100.5%
Grade FCC
Packaging Poly Bottle
Chemical Name or Material Sodium Metabisulfite, Granular