missing translation for 'onlineSavingsMsg'
Learn More

Sodium metabisulfite, Analytical Standard, ≥98.0% (iodometric, RT), MilliporeSigma™ Supelco™

Catalog No. 111015469
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015469 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015469 Supplier Merck Emd Millipore Supplier No. 089821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium metabisulfite is primarily used as an antioxidant and antimicrobial agent in a wide variety of foods and drugs. It is also used as the main additive in the prevention of melanosis in shrimp.

Specifications

CAS 7681-57-4
Linear Formula Na2S2O5
MDL Number MFCD00167602
Melting Point >300°C
Synonym E223; Sodium disulfite; Sodium pyrosulfite
Quantity 1 g
Formula Weight 190.11
Percent Purity ≥98.0% (iodometric, RT)
Grade Analytical Standard