Learn More
Sodium metabisulfite, ACS, 97% min
Description
Sodium metabisulfite is used as an active ingredient in campden tablets, as a cleaning agent for potable water reverse osmosis membranes, as a preservative and an antioxidant in food. It also acts as a corrosion inhibitor in oil and gas industry, as a bleaching agent in the production of coconut cream, as the source of sulfur dioxide and in the destruction of cyanide in commercial gold cyanidation processes. It is utilized in the precipitation of gold from auric acid as well as in the waste water treatment to remove hexavaent chromium as trivalent chromium by precipitation after reduction. Further, it is used in photography. In addition, it plays as an oxygen scavenger to eliminate the dissolved oxygen in wastewater and in pipes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >300°C |
| Color | White to Yellow |
| Physical Form | Crystalline Powder |
| Quantity | 2.5 kg |
| Odor | Pungent |
| Sensitivity | Air and moisture sensitive |
| Appearance | White to pale yellow powder/flake/crystalline/beads |
| Assay Percent Range | 97% min. |
| Density | 1.48 |
| Solubility Information | Soluble in water,ethanol and glycerol. |
| Show More |
Safety and Handling
| Hazard Category | H302-H318 |
| Precautionary Statement | P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c |
| EINECSNumber | 231-673-0 |
| RTECSNumber | UX8225000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only