Learn More
Sodium metabisulfite, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Sodium metabisulfite is used as an active ingredient in campden tablets, as a cleaning agent for potable water reverse osmosis membranes, as a preservative and an antioxidant in food. It also acts as a corrosion inhibitor in oil and gas industry, as a bleaching agent in the production of coconut cream, as the source of sulfur dioxide and in the destruction of cyanide in commercial gold cyanidation processes. It is utilized in the precipitation of gold from auric acid as well as in the waste water treatment to remove hexavaent chromium as trivalent chromium by precipitation after reduction. Further, it is used in photography. In addition, it plays as an oxygen scavenger to eliminate the dissolved oxygen in wastewater and in pipes.
Solubility
Soluble in water, ethanol and glycerol.
Notes
Moisture and air sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications
Specifications
| Melting Point | >300°C |
| pH | 3.5 to 5.0 |
| Physical Form | Crystalline Powder and/or Crystals |
| Quantity | 10,000 g |
| Odor | Like Rotten Eggs |
| Sensitivity | Air and moisture sensitive |
| Density | 1.48 |
| Linear Formula | Na2S2O5 |
| Merck Index | 14,8638 |
| Solubility Information | Soluble in water,ethanol and glycerol. |
| Show More |
Safety and Handling
| Hazard Category | H302-H318 |
| Hazard Statement | GHS H Statement H318-H302 Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c |
| EINECSNumber | 231-673-0 |
| RTECSNumber | UX8225000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only