missing translation for 'onlineSavingsMsg'
Learn More

Sodium meta-Bisulfite, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02004167
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004167 2.5 kg Wide Mouth Amber Glass Bottle
02004166 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02004167 Supplier Avantor Supplier No. 355205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Physical Form Granules
Quantity 2.5 kg
Formula Weight 190.11
Percent Purity ≥97.0%
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.48kg
Chemical Name or Material Sodium meta-Bisulfite