missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Iodide, 99%, Spectrum™ Chemical
$303.31 - $47553.72
Chemical Identifiers
| CAS | 7681-82-5 |
|---|---|
| Molecular Formula | INa |
| Molecular Weight (g/mol) | 149.89 |
| InChI Key | FVAUCKIRQBBSSJ-UHFFFAOYSA-M |
| IUPAC Name | sodium iodide |
| SMILES | [Na+].[I-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18613689
|
Spectrum Chemical Mfg Cor
S133025GM |
25 g | Amber Glass Bottle |
Each for $303.31
|
|
||||
|
18613691
|
Spectrum Chemical Mfg Cor
S1330500GM |
500 g | Amber Glass Bottle |
Each for $1,757.44
|
|
||||
|
18613688
|
Spectrum Chemical Mfg Cor
S133025KG |
2.5 kg | Amber Glass Bottle |
Each for $5,301.31
|
|
||||
|
18613686
|
Spectrum Chemical Mfg Cor
S133012KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18613690
|
Spectrum Chemical Mfg Cor
S133045KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.