missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide, 99%, Spectrum™ Chemical

Catalog No. 18613689
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613689 25 g Amber Glass Bottle
18613691 500 g Amber Glass Bottle
18613688 2.5 kg Amber Glass Bottle
18613686 12 kg Poly Pail
18613690 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613689 Supplier Spectrum Chemical Mfg Cor Supplier No. S133025GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.89
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
IUPAC Name sodium iodide
SMILES [Na+].[I-]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.