missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S133025KG

 View more versions of this product

Catalog No. 18613688


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7681-82-5
2.5 kg
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide
99%
Amber Glass Bottle
1
INa
[Na+].[I-]
149.89
Reagent

Chemical Identifiers

7681-82-5
149.89
sodium iodide
INa
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
[Na+].[I-]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.