missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide, 99%, Spectrum™ Chemical

Catalog No. 18613691
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613691 500 g Poly Pail
18613689 25 g Poly Pail
18613688 2.5 kg Poly Pail
18613686 12 kg Poly Pail
18613690 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613691 Supplier Spectrum Chemical Mfg Cor Supplier No. S1330500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.89
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
IUPAC Name sodium iodide
SMILES [Na+].[I-]

Specifications

CAS Max % 1
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Percent Purity 99%
Grade Reagent
Packaging Poly Pail
Chemical Name or Material Sodium Iodide

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.