missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide, ≥99%, (ACS Reagent Grade), MP Biomedicals

Catalog No. ICN19397990
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19397990 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19397990 Supplier MP Biomedicals Inc Supplier No. 0219397990

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range ≥99%
Formula Weight 149.9
Percent Purity 99.72%
Grade ACS Reagent
Chemical Name or Material Sodium Iodide
Recommended Storage Room Temperature