missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxymethanesulfinate Dihydrate 97.0+%, TCI America™

Catalog No. R0053500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0053500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R0053500G Supplier TCI America Supplier No. R0053500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 149-44-0
Molecular Formula CH3NaO3S
Molecular Weight (g/mol) 118.08
MDL Number MFCD00040426
InChI Key XWGJFPHUCFXLBL-UHFFFAOYSA-M
Synonym sodium hydroxymethanesulfinate, rongalite, rongalit, aldanil, discolite, formapon, formopan, hydrolit, leptacid, leptacit
PubChem CID 23689980
IUPAC Name sodium hydroxymethanesulfinate
SMILES [Na+].OCS([O-])=O

Specifications

Color White
Quantity 500 g
Formula Weight 118.08
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Hydroxymethanesulfinate Dihydrate
EINECSNumber (2)-0499&(2)-1633
RTECSNumber PB0380000
TSCA Yes