missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solution d’hydroxyde de sodium (50% en mousse), Fisher Chemical™
Spécifications
Spécifications
| pH | 12.0 |
| Nom ou substance chimique | Sodium Hydroxide Solution |
| Nom Remarque | 50% (w/w) |
| Point d’ébullition | 141,7 °C |
| Point de fusion | 5 °C |
| Fourchette de pourcentages d’essai | 50-52% |
| Quantité | 4 L |
| Désignation | Test Réussi |
| Emballage | Bouteille en polyéthylène |
| Pression de vapeur | 3 mmHg à 37 °C |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Causes severe burns by all exposure routes. Wear personal protective equipment. Ensure adequate ventilation. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. NFPA Health:3 Flammability:0 Instability:1 |
| DOTInformation | DOT Class 8, : Corrosive |