missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyde de sodium, dans le méthanol, certifié, 0,0100N ±0,0001N (0,01M), LabChem™

Numéro de catalogue. LC245202
Modifier l'affichage
Click to view available options
Quantité:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
LC245202 1 L
LC245204 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. LC245202 Fournisseur LabChem Code fournisseur LC245202

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Identifiants chimiques

CAS 1310-73-2 , 67-56-1
Formule moléculaire HNaO
Poids moléculaire (g/mol) 39.997
Clé InChI HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonyme sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
Nom de l’IUPAC Hydroxyde de sodium
SOURIRES [OH-].[Na+]

Spécifications

Nom ou substance chimique Sodium Hydroxide
Nom Remarque dans le méthanol
Couleur Incolore
CAS % maximal 99.96, 0.04
Formule linéaire NaOH
Concentration 0,0100N ±0,0001N (0,01 M)
Numéro UN UN1993
Renseignements sur la solubilité Soluble dans l’eau
Quantité 1 L
Poids de formule 40
Niveau Certifié
Désignation Test de réussite
Emballage Bouteille en polyéthylène
Traçabilité NIST Tracable à NIST
Forme physique Liquide
Afficher plus de résultats Afficher moins de résultats
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).
Precautionary Statement GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
In case of fire: Use alcohol resistant foam, extinguishing powder, sand to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Température ambiante