missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Dilution Solution, 0.16% (w/v), Ricca Chemical
Specifications
Specifications
| Quantity | 1 L |
| Percent Purity | 0.16% |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 0.16% (w/v) Aqueous Solution |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Sodium Hydroxide Dilution Solution |