missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyde de sodium, grade ACS, > 97,0%, LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Sodium Hydroxide |
| Couleur | Blanc |
| Point d’ébullition | 1 388 °C |
| Point de fusion | 323 °C |
| CAS % maximal | 100 |
| Formule linéaire | NaOH |
| Numéro UN | UN1823 |
| Densité | 2 130 kg/m3 |
| Renseignements sur la solubilité | Soluble dans l’eau |
| Quantité | 500 g |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Harmful in contact with skin. Causes severe skin burns and eye damage. Harmful to aquatic life. |
| Precautionary Statement | GHS P Statement Do not breathe dust, vapors. Wear eye protection, face protection, protective clothing, protective gloves. Wash exposed skin thoroughly after handling. Avoid release to the environment. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with applicable regulations. |
| EINECSNumber | 215-185-5 |
| Recommended Storage | Température ambiante |