missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, ≥98%, Honeywell

Numéro de catalogue. 60048166
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
60048166 6 x 1 kg
6001422 500 g
6001421 1 kg
6001423 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 60048166 Fournisseur Honeywell Chemicals Code fournisseur S58816X1KG
Il en reste null

Reagent Grade, pellets (anhydrous),≥98%

  • ≥95% for general laboratory use.

Identifiants chimiques

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Spécifications

Linear Formula NaOH
UN Number UN1823
Quantity 6 x 1 kg
Formula Weight 40
Percent Purity ≥98%
Vapor Density >1 (vs air)
Vapor Pressure 3 mmHg (37°C), 18 mmHg (20°C)
Chemical Name or Material Sodium hydroxide
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
DOTInformation 8 / PGII
RTECSNumber 215-185-5