missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 50% (w/v) Solution, Spectrum™ Chemical
Spécifications
Spécifications
| Quantity | 500 mL |
| Percent Purity | 50% ±1% |
| Grade | Chemical Solution |
| Packaging | Poly Bottle |
| Chemical Name or Material | Sodium Hydroxide Solution |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.