missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 30% (w/v), Ricca Chemical

Catalog No. 72741
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
72741 4 L Natural Poly Bottle
727416 500 mL Natural Poly Bottle
727432 1 L Natural Poly Bottle
72745 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 72741 Supplier Ricca Chemical Company Supplier No. 72741
Edge

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Min % 75.0, 22.69
CAS Max % 78.0, 23.61
Percent Purity 30%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH >13
Quantity 4 L
Concentration or Composition Notes 30% (w/v)
Chemical Name or Material Sodium Hydroxide, 30% (w/v)