missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 3.00 N, Ricca Chemical

Catalog No. p-3252361
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
745916 500 mL Natural Poly Bottle
745932 1 L Natural Poly Bottle
74591 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 745916 Supplier Ricca Chemical Company Supplier No. 745916
Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Min % 79.0, 19.03
CAS Max % 82.0, 19.8
Grade Laboratory
Packaging Natural Poly Bottle
pH >13
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 3.00Normal
Chemical Name or Material Sodium Hydroxide