missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 24% (w/v), Ricca Chemical
Spécifications
Spécifications
| Color | Colorless |
| CAS Min % | 67.0, 31.45 |
| CAS Max % | 69.0, 32.73 |
| Quantity | 500 mL |
| Percent Purity | 23.94 to 24.06% |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 24% (w/v) Aqueous Solution |
| Packaging | Natural Poly Bottle |
| Physical Form | Liquid |
| Chemical Name or Material | Sodium Hydroxide |
Sécurité et manipulation
| missing translation for 'shelfLife' | 24 months |