missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 2.00 N, Ricca Chemical

Numéro de catalogue. p-3252355
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
745516 500 mL Natural Poly Bottle
745532 1 L Natural Poly Bottle
R74550004F 4 L Cubitainer
74551 4 L Natural Poly Bottle
745525 10 L Cubitainer
74555 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. 745516 Fournisseur Ricca Chemical Company Code fournisseur 745516
Il en reste null

Identifiants chimiques

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Spécifications

Color Colorless
CAS Min % 91.0, 7.4
CAS Max % 94.0, 7.7
Concentration or Composition Notes 2.00Normal
Quantity 500 mL
Grade Laboratory
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide