missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide (1mol/L in Water), TCI America™
Spécifications
Spécifications
| CAS | 1310-73-2 |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Molecular Weight (g/mol) | 39.997 |