missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 1% (w/v) (10 g/L), Ricca Chemical
Supplier: Ricca Chemical Company 724332
Specifications
| Sodium Hydroxide Solution | |
| 97.0,0.97 | |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 1% | |
| Natural Poly Bottle | |
| >13 | |
| 1% (w/v),10g/L |
| 1310-73-2,7732-18-5 | |
| 100.0,1.01 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| Laboratory | |
| Aqueous Solution | |
| 1 L |
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |