Learn More
Andwin Scientific Inc Sodium hydroxide 1.0 N | 1310-73-2 | MFCD00003548 | 1L
Numéro de catalogue. NC0602989
Encompass
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH Sodium Hydroxide is an odorless liquid chemical. It has a Health Hazard NFPA Rating of 3, a Fire Hazard NFPA Rating of 0, and a Reactivity NFPA Rating of 0. This chemical complies with the APHA standard, AOAC 936.16 standard, and ASTM E200-91 standard. It has a shelf life of 2 years. This product is packaged and sold in a 1 L poly bottle.
Numéro de catalogue. NC0602989 Fournisseur Andwin Scientific Inc Code fournisseur BDH72221
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH Sodium Hydroxide is an odorless liquid chemical. It has a Health Hazard NFPA Rating of 3, a Fire Hazard NFPA Rating of 0, and a Reactivity NFPA Rating of 0. This chemical complies with the APHA standard, AOAC 936.16 standard, and ASTM E200-91 standard. It has a shelf life of 2 years. This product is packaged and sold in a 1 L poly bottle.