missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.600 N, Ricca Chemical
Spécifications
Spécifications
| Nom ou substance chimique | Sodium Hydroxide |
| CAS | 7732-18-6 |
| Quantité | 20 L |
| Concentration ou composition (par analyte ou composants) | 0.600N |
| Emballage | Cubitainer |