missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.4% (w/v) (4 g/L), Ricca Chemical

Supplier:  Ricca Chemical Company 7242516

 View more versions of this product

Catalog No. 7242-516

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide
98.0,0.39
HNaO
500 mL
[OH-].[Na+]
39.997
CHEBI:32145
Natural Poly Bottle
1310-73-2,7732-18-5
100.0,0.41
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
0.4% (w/v) Aqueous Solution (4g/L)

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS Documents

Documents

Promotions

Promotions