missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.313 N (12.5 g/L), Ricca Chemical

Catalog No. 737532
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
737532 1 L Natural Poly Bottle
73751 4 L Natural Poly Bottle
73755 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 737532 Supplier Ricca Chemical Company Supplier No. 737532
Only null left

Suitable for Crude Fiber Analysis of Food and Feed

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Min % 97.0, 1.24
CAS Max % 100.0, 1.29
Grade Laboratory
Packaging Natural Poly Bottle
pH >13
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.313Normal,12.5g/L
Chemical Name or Material Sodium Hydroxide, 0.313N (12.5g/L)