missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.250 N (N/4), Ricca Chemical

Catalog No. 737016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
737016 500 mL Natural Poly Bottle
737032 1 L Natural Poly Bottle
73701 4 L Natural Poly Bottle
737025 10 L Cubitainer
73705 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 737016 Supplier Ricca Chemical Company Supplier No. 737016

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Min % 96.0, 1.93
CAS Max % 100.0, 2.01
Grade Laboratory
Packaging Natural Poly Bottle
pH >13
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.250N
Chemical Name or Material Sodium Hydroxide