missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.250 N (N/4), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 96.0, 1.93 |
| CAS Max % | 100.0, 2.01 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | >13 |
| Quantity | 4 L |
| Physical Form | Liquid |
| Concentration or Composition Notes | 0.250N |
| Chemical Name or Material | Sodium Hydroxide |