missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.0200 N (N/50), 1 mL = 1 mg CaCO3, Ricca Chemical
$68.53 - $1195.89
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 40 g/mol |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
730016
|
Ricca Chemical Company
RI730016 |
500 mL | Poly Natural Bottle |
Each for $68.53
|
|
||||
|
7300-32
|
Ricca Chemical Company
730032 |
1 L | Poly Natural Bottle |
Each for $94.63
|
|
||||
|
7300-1
|
Ricca Chemical Company
73001 |
4 L | Poly Natural Bottle |
Each for $215.87
|
|
||||
|
R730000020F
|
Ricca Chemical Company
R730000020F |
20 L | Cubitainer |
Each for $1,195.89
|
|
||||
Description
- Tested in an ISO 17025 Accredited Facility
- Test Reports provided with Certified Values and Measurement Uncertainty
- Tight release specifications for low lot-to-lot variability
Chemical Identifiers
| 1310-73-2 | |
| 40 g/mol | |
| 14798 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| CHEBI:32145 |
Specifications
| Colorless | |
| 98%,0.16 | |
| HNaO | |
| 0.0200 N | |
| 500 mL | |
| [OH-].[Na+] | |
| 40 g/mol | |
| CHEBI:32145 | |
| Poly Natural Bottle | |
| Sodium Hydroxide |
| 1310-73-2,7732-18-5 | |
| 100%,0.16 | |
| 0.01995 to 0.02005 N | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| Laboratory | |
| Liquid | |
| Normal at 20°C |
Safety and Handling
ShelfLife : 24 months