missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.0200 N (N/50), 1 mL = 1 mg CaCO3, Ricca Chemical

Supplier:  Ricca Chemical Company 73001

 View more versions of this product

Catalog No. 7300-1


Only null left
Explore available promotions

Description

Description

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability
Specifications

Specifications

Sodium Hydroxide
1310-73-2,7732-18-5
100%,0.16
HNaO
0.0200 N
4 L
[OH-].[Na+]
40 g/mol
CHEBI:32145
Poly Natural Bottle
Colorless
98%,0.16
Normal at 20°C
0.01995 to 0.02005 N
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Laboratory
Liquid

Chemical Identifiers

1310-73-2
40 g/mol
14798
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
CHEBI:32145
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
Documents

Documents

Promotions

Promotions