missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.0200 N (N/50) in Methanol, Ricca Chemical
$188.97 - $188.97
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | CH4O |
| Molecular Weight (g/mol) | 32.04 |
| MDL Number | MFCD00004595 |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| IUPAC Name | methanol |
| SMILES | CO |
Chemical Identifiers
| 1310-73-2 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CO |
Specifications
| Colorless | |
| 0.19,95.0,2.35 | |
| CH4O | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CO | |
| 32.04 | |
| Natural Poly Bottle | |
| >12 | |
| Liquid | |
| Sodium Hydroxide |