missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen arsenate heptahydrate, ACS, 98.0-102.0%

Catalog No. AA3337314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3337314 25 g
AA3337322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3337314 Supplier Thermo Scientific Chemicals Supplier No. 3337314
Only null left

CAS: 10048-95-0 | AsH15Na2O11 | 312.01 g/mol

Sodium hydrogen arsenate heptahydrate is used as a source of soluble arsenic and in dyeing and printing with Turkey-red oil. It also acts as a veterinary drug for anti-parasitic, skin diseases, blood diseases, rheumatism and asthma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10048-95-0
Molecular Formula AsH15Na2O11
Molecular Weight (g/mol) 312.01
MDL Number MFCD00149179
InChI Key KOLXPEJIBITWIQ-UHFFFAOYSA-L
Synonym disodium arsenate heptahydrate, sodium acid arsenate, heptahydrate, sodium arsenate heptahydrate, sodium orthoarsenate heptahydrate, dibasic sodium arsenate heptahydrate, sodium arsenate, dibasic, heptahydrate, unii-90zq7g407w, disodium arsenate, heptahydrate, ccris 5553, arsenic acid h3aso4 , disodium salt, heptahydrate
PubChem CID 61460
SMILES O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O

Specifications

Melting Point 130°C (decomposition)
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in water and glycerol.
IUPAC Name disodium heptahydrate hydrogen arsorate
Formula Weight 312.01 (185.94 Anhydrous)
Grade ACS
Appearance White granular
Density 1.88 g/cm3 at 20°C
Chemical Name or Material Sodium hydrogen arsenate heptahydrate, For analysis ACS
Show More Show Less
Hazard Category H301+H331-H350
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation DOT Class: 6.1; Packing Group: II
EINECSNumber 231-902-4
RTECSNumber CG0900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only