missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydride (60%, dispersion in Paraffin Liquid), TCI America™

Catalog No. S0481100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0481100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0481100G Supplier TCI America Supplier No. S0481100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7646-69-7
Molecular Formula HNa
Molecular Weight (g/mol) 23.998
MDL Number MFCD00003471
InChI Key BZKBCQXYZZXSCO-UHFFFAOYSA-N
Synonym sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid
PubChem CID 24758
IUPAC Name sodium;hydride
SMILES [H-].[Na+]

Specifications

Physical Form Crystalline Powder
Quantity 100 g
UN Number 1427
Formula Weight 24.00
Chemical Name or Material Sodium Hydride (60%, dispersion in Paraffin Liquid)
EINECSNumber (1)-0409
RTECSNumber WB3910000
TSCA Yes