missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexametaphosphate, Thermo Scientific Chemicals

Catalog No. AC612175000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC612175000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC612175000 Supplier Thermo Scientific Chemicals Supplier No. 612175000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68915-31-1
Molecular Formula H2Na2O7P2
Molecular Weight (g/mol) 221.94
MDL Number MFCD00136045
InChI Key GYQBBRRVRKFJRG-UHFFFAOYSA-L
IUPAC Name disodium hydrogen (hydrogen phosphonatooxy)phosphonate
SMILES [Na+].[Na+].OP([O-])(=O)OP(O)([O-])=O

Specifications

Physical Form Powder
Quantity 500 g
Linear Formula Na6(PO3)6
Formula Weight 611.52
Percent Purity 98%
Grade Ungraded
Identification Pass Test
Density 2.1810g/mL
Chemical Name or Material Sodium hexametaphosphate

RUO – Research Use Only