missing translation for 'onlineSavingsMsg'
Learn More

Sodium Glycolate, TCI America™

Catalog No. G0111500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0111500G 500 g
G011125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0111500G Supplier TCI America Supplier No. G0111500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2836-32-0
Molecular Formula C2H3NaO3
Molecular Weight (g/mol) 98.033
MDL Number MFCD00065358
InChI Key VILMUCRZVVVJCA-UHFFFAOYSA-M
Synonym sodium glycolate, sodium hydroxyacetate, sodium 2-hydroxyacetate, sodium lycolate, monosodium glycolate, glycolic acid sodium salt, natriumglykolat, sodium glycollate, acetic acid, hydroxy-, monosodium salt, glycolic acid, monosodium salt
PubChem CID 517347
IUPAC Name sodium;2-hydroxyacetate
SMILES C(C(=O)[O-])O.[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 98.03
Physical Form Crystalline Powder
Chemical Name or Material Sodium Glycolate
EINECSNumber (2)-3057
RTECSNumber MC5525000
TSCA Yes