missing translation for 'onlineSavingsMsg'
Learn More

Sodium glycolate, 97%

Catalog No. AAA1234136
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1234136 500 g
AAA1234122 100 g
AAA123410E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1234136 Supplier Thermo Scientific Chemicals Supplier No. A1234136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 2836-32-0 | C2H3NaO3 | 98.033 g/mol

Sodium glycolate is used as a disintegrant, suspending agent, gelling agent, buffering agent and in pharmaceutical grade for tablets and capsules. Further, it is used in cosmetics and personal care products primarily as an exfoliant, as pH adjusters, skin-conditioning agents and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium glycolate is used as a disintegrant, suspending agent, gelling agent, buffering agent and in pharmaceutical grade for tablets and capsules. Further, it is used in cosmetics and personal care products primarily as an exfoliant, as pH adjusters, skin-conditioning agents and as a flavoring agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 2836-32-0
Molecular Formula C2H3NaO3
Molecular Weight (g/mol) 98.033
MDL Number MFCD00065358
InChI Key VILMUCRZVVVJCA-UHFFFAOYSA-M
Synonym sodium glycolate, sodium hydroxyacetate, sodium 2-hydroxyacetate, sodium lycolate, monosodium glycolate, glycolic acid sodium salt, natriumglykolat, sodium glycollate, acetic acid, hydroxy-, monosodium salt, glycolic acid, monosodium salt
PubChem CID 517347
IUPAC Name sodium;2-hydroxyacetate
SMILES C(C(=O)[O-])O.[Na+]

Specifications

Melting Point 210°C to 218°C
Assay Percent Range 97%
Linear Formula HOCH2CO2Na
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 98.03
Percent Purity 97%
Physical Form Powder or Lumps
Chemical Name or Material Sodium glycolate
EINECSNumber 220-624-9
RTECSNumber MC5525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only